Merck
Phospholipid-bound molecular rotors: synthesis and characterization.
Bioorganic & medicinal chemistry (2002-09-06)
Mark A Haidekker, Thomas Brady, Ke Wen, Cliff Okada, Hazel Y Stevens, Jeniffer M Snell, John A Frangos, Emmanuel A Theodorakis
ABSTRACT
Molecular rotors are fluorescent molecules with a viscosity-sensitive quantum yield that are often used to measure viscosity changes in cell membranes and liposomes. However, commercially available molecular rotors, such as DCVJ (1) do not localize in cell membranes but rapidly migrate into the cytoplasm leading to unreliable measurements of cell membrane viscosity. To overcome this problem, we synthesized molecular rotors covalently attached to a phospholipid scaffold. Attaching the rotor group to the hydrophobic end of phosphatidylcholine (PC) did not affect the rotor's viscosity sensitivity and allowed adequate integration into artificial bilayers as well as complete localization in the plasma membrane of an endothelial cell line. Moreover, these new rotors enabled the monitoring of phospholipid transition temperature. However, attachment of the rotor groups to the hydrophilic head of the phospholipid led to a partial loss of viscosity sensitivity. The improved sensitivity and exclusive localization in the cell plasma membrane exhibited by the phospholipid-bound molecular rotors suggest that these probes can be used for the study of membrane microviscosity.
Sigma-Aldrich
9-(2,2-Dicyanovinyl)julolidine, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)