MilliporeSigma
  • alpha,beta-poly(asparthylhydrazide)-glycidyltrimethylammonium chloride copolymers (PAHy-GTA): novel polymers with potential for DNA delivery.
alpha,beta-poly(asparthylhydrazide)-glycidyltrimethylammonium chloride copolymers (PAHy-GTA): novel polymers with potential for DNA delivery.
Journal of controlled release : official journal of the Controlled Release Society (2001-11-02)
E Pedone, G Cavallaro, S C Richardson, R Duncan, G Giammona
ABSTRACT
Hydrophilic polycations form complexes when mixed with plasmids. Following functionalisation with glycidyltrimethylammonium chloride (GTA) alpha,beta-poly(asparthylhydrazide) (PAHy), a water-soluble synthetic macromolecule, becomes polycationic and potentially useful for systemic gene delivery. Initially the biocompatibility of PAHy and PAHy-GTA derivatives with different degrees of positive charge substitution were studied and it was shown that PAHy-GTA was neither haemolytic nor cytotoxicity up to 1 mg/ml. After intravenous injection (125)I-labelled PAHy-GTA derivative containing 46 mol% (PAHy-GTA(b)) of trimethylammonium groups did not accumulate in the liver (4.1+/-0.9% of the recovered dose after 1 h) but was subjected to renal excretion (45+/-21% of the recovered dose was in the kidneys after 1 h). PAHy-GTA formed complexes with DNA (gel retardation) and they protected against degradation by DNase II. Finally the ability of the PAHy-GTA(b) derivative to mediate the transfection of HepG2 cells using the marker gene beta-galactosidase was studied. The optimum plasmid/polymer mass ratio was examined in comparison to LipofectACE, Lipofectin and polyethylenimine.
Sigma-Aldrich
Glycidyltrimethylammonium chloride, technical, ≥90% (calc. based on dry substance, AT)