MilliporeSigma
  • RNA polymerase inhibitors with activity against rifampin-resistant mutants of Staphylococcus aureus.
RNA polymerase inhibitors with activity against rifampin-resistant mutants of Staphylococcus aureus.
Antimicrobial agents and chemotherapy (2000-10-19)
A O'Neill, B Oliva, C Storey, A Hoyle, C Fishwick, I Chopra
ABSTRACT
A collection of rifampin-resistant mutants of Staphylococcus aureus with characterized RNA polymerase beta-subunit (rpoB) gene mutations was cross-screened against a number of other RNA polymerase inhibitors to correlate susceptibility with specific rpoB genotypes. The rpoB mutants were cross-resistant to streptolydigin and sorangicin A. In contrast, thiolutin, holomycin, corallopyronin A, and ripostatin A retained activity against the rpoB mutants. The second group of inhibitors may be of interest as drug development candidates.
Sigma-Aldrich
Thiolutin, from Streptomyces luteosporeus, ≥95% (HPLC)