MilliporeSigma
Protein disulfide isomerase mediates integrin-dependent adhesion.
FEBS letters (2000-06-20)
J Lahav, N Gofer-Dadosh, J Luboshitz, O Hess, M Shaklai
ABSTRACT
Cell adhesion is mediated by the integrin adhesion receptors. Receptor-ligand interaction involves conformational changes in the receptor, but the underlying mechanism remains unclear. Our earlier work implied a role for sulfhydryls in integrin response to ligand binding in the intact blood platelet. We now show that non-penetrating blockers of free sulfhydryls inhibit beta(1) and beta(3) integrin-mediated platelet adhesion regardless of the affinity state of the integrin. Removal of the inhibitors prior to adhesion fully restores adhesion despite the irreversible nature of inhibitor-thiol interaction, indicating sulfhydryl exposure in response to adhesion. We further show that blocking protein disulfide isomerase (PDI) inhibits adhesion. These data indicate that: (a) ecto-sulfhydryls are necessary for integrin-mediated platelet adhesion; (b) disulfide exchange takes place during this process; (c) surface PDI is involved in integrin-mediated adhesion.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Monobromo(trimethylammonio)bimane bromide, suitable for fluorescence, ≥90.0% (HPLC)