MilliporeSigma
  • Formation of hyodeoxycholic acid from muricholic acid and hyocholic acid by an unidentified gram-positive rod termed HDCA-1 isolated from rat intestinal microflora.
Formation of hyodeoxycholic acid from muricholic acid and hyocholic acid by an unidentified gram-positive rod termed HDCA-1 isolated from rat intestinal microflora.
Applied and environmental microbiology (1999-07-02)
H J Eyssen, G De Pauw, J Van Eldere
ABSTRACT
From the rat intestinal microflora we isolated a gram-positive rod, termed HDCA-1, that is a member of a not previously described genomic species and that is able to transform the 3alpha,6beta, 7beta-trihydroxy bile acid beta-muricholic acid into hyodeoxycholic acid (3alpha,6alpha-dihydroxy acid) by dehydroxylation of the 7beta-hydroxy group and epimerization of the 6beta-hydroxy group into a 6alpha-hydroxy group. Other bile acids that were also transformed into hyodeoxycholic acid were hyocholic acid (3alpha, 6alpha,7alpha-trihydroxy acid), alpha-muricholic acid (3alpha,6beta, 7alpha-trihydroxy acid), and omega-muricholic acid (3alpha,6alpha, 7beta-trihydroxy acid). The strain HDCA-1 could not be grown unless a nonconjugated 7-hydroxylated bile acid and an unidentified growth factor produced by a Ruminococcus productus strain that was also isolated from the intestinal microflora were added to the culture medium. Germfree rats selectively associated with the strain HDCA-1 plus a bile acid-deconjugating strain and the growth factor-producing R. productus strain converted beta-muricholic acid almost completely into hyodeoxycholic acid.
Sigma-Aldrich
Hyodeoxycholic acid, ≥98%