MilliporeSigma
  • Ras-dependent induction of cellular responses by constitutively active phosphatidylinositol-3 kinase.
Ras-dependent induction of cellular responses by constitutively active phosphatidylinositol-3 kinase.
Science (New York, N.Y.) (1995-04-07)
Q Hu, A Klippel, A J Muslin, W J Fantl, L T Williams
ABSTRACT
Phosphatidylinositol (Pl)-3 kinase is one of many enzymes stimulated by growth factors. A constitutively activated mutant, p110, that functions independently of growth factor stimulation was constructed to determine the specific responses regulated by Pl-3 kinase. The p110 protein exhibited high specific activity as a Pl-3 kinase and as a protein kinase. Expression of p110 in NIH 3T3 cells induced transcription from the fos promoter. Co-expression of dominant negative Ras blocked this response. When expressed in Xenopus laevis oocytes, p110 increased the amount of guanosine 5'-triphosphate-bound Ras, caused activation of the Ras effector Raf-1, and induced Ras-dependent oocyte maturation. These findings show that Pl-3 kinase can stimulate diverse Ras-dependent cellular processes, including oocyte maturation and fos transcription.