MilliporeSigma
  • Studies on a new proteolytic enzyme from A chromobacter lyticus M497-1. I. Purification and some enzymatic properties.
Studies on a new proteolytic enzyme from A chromobacter lyticus M497-1. I. Purification and some enzymatic properties.
Biochimica et biophysica acta (1981-07-24)
T Masaki, M Tanabe, K Nakamura, M Soejima
ABSTRACT
Achromobacter lyticus M497-1 produces three kinds of alkaline proteases (protease I, II and III) in culture medium along with the bacteriolytic enzyme (Masaki, T., Nakamura, K., Isono, M. and Soejima, M. (1978) Agric. Biol. Chem. 42, 1443--1445). Among these three proteases, Achromobacter protease I (EC 3.4.21.-) shows strict splitting for lysine residues at the carboxyl side of the splitting point. This enzyme was purified through a sequence of benzalkonium chloride treatment, acetone fractionation, CM-cellulose and DEAE-cellulose treatment chromatography on AH-Sepharose 4B and isoelectric focusing method. This form was shown to be homogeneous by polyacrylamide gel electrophoresis and ultracentrifugation analysis. The physicochemical properties of the enzyme were: Mr 30 500; partial specific volume (v), 0.717 ml/g; intrinsic viscosity (nu), 0.0385) dl/g; isoelectric point (pI) 6.9; and E1%1cm at 280 nm, 18.77. The enzyme was composed of 294 residues of amino acid per molecule, with glycine as NH2-terminal and lysine as COOH-terminal amino acids. The optimum pH values with casein, Bz-lys-pNA and Tos-Lys-OMe were 8.5--10.7, 9.0--9.5 and 7.8--8.2, respectively. The enzyme was inhibited by iPr2P-F, PhCH2SO2F and Tos-LysCH2Cl but not by Tos-ArgCH2Cl, EDTA, o-phenanthroline and PCMB.
Sigma-Aldrich
Achromopeptidase from bacteria, partially purified powder, ≥20,000 units/mg solid