MilliporeSigma
  • Qualitative and quantitative analysis of glycated proteins in human plasma by glucose isotopic labeling with ¹³C6-reducing sugars.
Qualitative and quantitative analysis of glycated proteins in human plasma by glucose isotopic labeling with ¹³C6-reducing sugars.
Methods in molecular biology (Clifton, N.J.) (2011-04-07)
Feliciano Priego-Capote, Maria Ramírez-Boo, Denis Hochstrasser, Jean-Charles Sanchez
ABSTRACT
Glucose is the predominant source of energy in cells. However, a chronic high glucose exposure of proteins modifies a number of biological pathways, known as glucotoxicity. Several studies have suggested that this impaired protein function is associated in part to protein glycation. However, despite the evidence of this glucotoxicity on tissues and cells, the exact mechanisms underlying the loss of protein function by glycation are not well understood. Strategies that will allow the discovery of the identity and function of the glycated plasma proteins generated by chronic hyperglycemia would be of a considerable help to further understand the underlying mechanisms implicated in protein dysfunction associated with glucotoxicity. The present chapter describes an innovative and interdisciplinary proteomics strategy to achieve a comprehensive identification and quantification of glycated proteins in plasma. This should provide new molecular insights into protein glycation mechanisms and identify new targets to improve the subsequent defective protein function.
Sigma-Aldrich
Endoproteinase Glu-C from Staphylococcus aureus V8, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Endoproteinase Glu-C from Staphylococcus aureus V8, Type XVII-B, lyophilized powder, 500-1,000 units/mg solid
Sigma-Aldrich
Plasma, from human