MilliporeSigma
  • Mixed glucose and lactate uptake by Corynebacterium glutamicum through metabolic engineering.
Mixed glucose and lactate uptake by Corynebacterium glutamicum through metabolic engineering.
Biotechnology journal (2011-03-04)
Andreas Neuner, Elmar Heinzle
ABSTRACT
The Corynebacterium glutamicum ATCC 13032 lysC(fbr) strain was engineered to grow fast on racemic mixtures of lactate and to secrete lysine during growth on lactate as well as on mixtures of lactate and glucose. The wild-type C. glutamicum only grows well on L-lactate. Overexpression of D-lactate dehydrogenase (dld) achieved by exchanging the native promoter of the dld gene for the stronger promoter of the sod gene encoding superoxide dismutase in C. glutamicum resulted in a duplication of biomass yield and faster growth without any secretion of lysine. Elementary mode analysis was applied to identify potential targets for lysine production from lactate as well as from mixtures of lactate and glucose. Two targets for overexpression were pyruvate carboxylase and malic enzyme. The overexpression of these genes using again the sod promoter resulted in growth-associated production of lysine with lactate as sole carbon source with a carbon yield of 9% and a yield of 15% during growth on a lactate-glucose mixture. Both substrates were taken up simultaneously with a slight preference for lactate. As surmised from the elementary mode analysis, deletion of glucose-6-phosphate isomerase resulted in a decreased production of lysine on the mixed substrate. Elementary mode analysis together with suitable objective functions has been found a very useful tool guiding the design of strains producing lysine on mixed substrates.
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, ammonium sulfate suspension, ≥250 units/mg protein (biuret)
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, lyophilized powder, 150-500 units/mg protein
Sigma-Aldrich
D-Lactic Dehydrogenase from Staphylococcus epidermidis, lyophilized powder, ≥80 units/mg solid