MilliporeSigma
  • Efficient production of (R)-o-chloromandelic acid by deracemization of o-chloromandelonitrile with a new nitrilase mined from Labrenzia aggregata.
Efficient production of (R)-o-chloromandelic acid by deracemization of o-chloromandelonitrile with a new nitrilase mined from Labrenzia aggregata.
Applied microbiology and biotechnology (2012-03-29)
Chen-Sheng Zhang, Zhi-Jun Zhang, Chun-Xiu Li, Hui-Lei Yu, Gao-Wei Zheng, Jian-He Xu
ABSTRACT
(R)-o-Chloromandelic acid is the key precursor for the synthesis of Clopidogrel®, a best-selling cardiovascular drug. Although nitrilases are often used as an efficient tool in the production of α-hydroxy acids, there is no practical nitrilase specifically developed for (R)-o-chloromandelic acid. In this work, a new nitrilase from Labrenzia aggregata (LaN) was discovered for the first time by genomic data mining, which hydrolyzed o-chloromandelonitrile with high enantioselectivity, yielding (R)-o-chloromandelic acid in 96.5% ee. The LaN was overexpressed in Escherichia coli BL21 (DE3), purified, and its catalytic properties were studied. When o-chloromandelonitrile was used as the substrate, the V(max) and K(m) of LaN were 2.53 μmol min⁻¹ mg⁻¹ protein and 0.39 mM, respectively, indicating its high catalytic efficiency. In addition, a study of substrate spectrum showed that LaN prefers to hydrolyze arylacetonitriles. To relieve the substrate inhibition and to improve the productivity of LaN, a biphasic system of toluene-water (1:9, v/v) was adopted, in which o-chloromandelonitrile of 300 mM (apparent concentration, based on total volume) could be transformed by LaN in 8 h, giving an isolated yield of 94.5%. The development of LaN makes it possible to produce (R)-o-chloromandelic acid by deracemizing o-chloromandelonitrile with good ee value and high substrate concentration.
Sigma-Aldrich
Nitrilase, recombinant, expressed in E. coli, ≥2.0 U/mg