MilliporeSigma
  • Quantification of hydrolysis of toxic organophosphates and organophosphonates by diisopropyl fluorophosphatase from Loligo vulgaris by in situ Fourier transform infrared spectroscopy.
Quantification of hydrolysis of toxic organophosphates and organophosphonates by diisopropyl fluorophosphatase from Loligo vulgaris by in situ Fourier transform infrared spectroscopy.
Analytical biochemistry (2008-12-17)
Jürgen Gäb, Marco Melzer, Kai Kehe, André Richardt, Marc-Michael Blum
ABSTRACT
The enzyme diisopropyl fluorophosphatase (DFPase) from the squid Loligo vulgaris effectively catalyzes the hydrolysis of diisopropyl fluorophosphate (DFP) and a number of organophosphorus nerve agents, including sarin, soman, cyclosarin, and tabun. Up to now, the determination of kinetic data has been achieved by techniques such as pH-stat titration, ion-selective electrodes, and fluorogenic substrate analogs. We report a new assaying method using in situ Fourier transform infrared (FTIR) spectroscopy with attenuated total reflection (ATR) for the real-time determination of reaction rates. The method employs changes in the P-O-R stretching vibration of DFP and nerve agent substrates when hydrolyzed to their corresponding phosphoric and phosphonic acids. It is shown that the Lambert-Beer law holds and that changes in absorbance can be directly related to changes in concentration. Compared with other methods, the use of in situ FTIR spectroscopy results in a substantially reduced reaction volume that adds extra work safety when handling highly toxic substrates. In addition, the new method allows the noninvasive measurement of buffered solutions with varying ionic strengths complementing existing methods. Because the assay is independent of the used enzyme, it should also be applicable to other phosphotriesterase enzymes such as organophosphorus hydrolase (OPH), organophosphorus acid anhydrolase (OPAA), and paraoxonase (PON).
Sigma-Aldrich
Diisopropyl-fluorophosphatase, ≥30 U/mg