MilliporeSigma
  • The transforming growth factor-beta system, a complex pattern of cross-reactive ligands and receptors.
The transforming growth factor-beta system, a complex pattern of cross-reactive ligands and receptors.
Cell (1987-02-13)
S Cheifetz, J A Weatherbee, M L Tsang, J K Anderson, J E Mole, R Lucas, J Massagué
ABSTRACT
A new homodimer form of transforming growth factor-beta (TGF-beta), TGF-beta 2, has been identified in porcine blood platelets. TGF-beta 2 is homologous to ordinary TGF-beta (TGF-beta 1), which is also present in platelets. TGF-beta 1.2, a heterodimer containing one TGF-beta 1 chain and one TGF-beta 2 chain, has also been isolated. TGF-beta 1 and TGF-beta 2 interact differently with a family of receptors in target cells. A 280 kd receptor displays high affinity for both TGF-beta 1 and TGF-beta 2. Occupancy of this receptor by TGF-beta 1 or TGF-beta 2 correlates with the ability of these TGF-beta s to inhibit cell proliferation. In contrast, 65 kd and 85 kd receptors have high affinity for TGF-beta 1 but lower affinity for TGF-beta 2. The existence of distinct forms of TGF-beta that interact differently with a family of TGF-beta receptors could provide flexibility to the regulation of tissue growth and differentiation by the TGF-beta system.