MilliporeSigma
  • Cloning of a novel component of A-type K+ channels operating at subthreshold potentials with unique expression in heart and brain.
Cloning of a novel component of A-type K+ channels operating at subthreshold potentials with unique expression in heart and brain.
Journal of neurophysiology (1996-05-01)
P Serôdio, E Vega-Saenz de Miera, B Rudy
ABSTRACT
1. Proteins of the Kv4 or Shal-related subfamily are key components of transient K+ channels (A channels) operating at subthreshold values of the membrane potential. We have cloned and characterized a new mammalian Kv4 or Shal-related cDNA (Kv4.3) that predicts a protein with strong sequence conservation with the other known members of this subfamily. 2. Injection of Kv4.3 transcripts into Xenopus oocytes generates an A type K+ current, with small but physiologically significant differences from the currents expressed by Kv4.2 and Kv4.1 mRNAs. Kv4.3 currents can be modified to resemble native A currents by coinjection with a low molecular weight mRNA fraction from rat brain which does not express detectable currents on its own. Particularly striking is a 7-to-10-fold increase in the rate of recovery from inactivation, a 5- to 10-fold increase in current magnitude and a 3- to 4-fold increase in sensitivity to 4-amino pyridine (4-AP). 3. In situ hybridization histochemistry was used to compare the expression of the three known Kv4 genes. Kv4.2 and Kv4.3 (but not Kv4.1) are abundant in the adult rat brain, with each displaying a specific, but sometimes overlapping pattern of expression. Moreover, a reciprocal gradient of expression of Kv4.2 and Kv4.3 transcripts is seen in some brain areas, such as in the pyramidal cell layers of the hippocampus and the granule cell layer of the cerebellum. Therefore Kv4 proteins may form heteromultimeric channels of distinct subunit composition in different neurons. Moreover, the results suggest that neurons such as pyramidal cells in the hippocampus and granule cells in the cerebellum represent heterogeneous cell populations in terms of their ISA, and hence in their firing patterns. Kv4.2 and Kv4.3 also display complementary expression in the heart, with Kv4.3 being more abundant in atria and Kv4.2 in ventricle. The existence of multiple Kv4 proteins forming channels of variable subunit combinations helps explain the diversity of ISA channels in neurons.