MilliporeSigma
  • Molecular cloning of multiple subtypes of a novel rat brain isoform of the alpha 1 subunit of the voltage-dependent calcium channel.
Molecular cloning of multiple subtypes of a novel rat brain isoform of the alpha 1 subunit of the voltage-dependent calcium channel.
Neuron (1991-07-01)
A Hui, P T Ellinor, O Krizanova, J J Wang, R J Diebold, A Schwartz
ABSTRACT
Several cDNAs encoding an isoform of the alpha 1 subunit of the voltage-dependent calcium channel were isolated from rat brain cDNA libraries. The complete nucleotide sequence of 6975 bp encodes a protein of 1634 amino acids, which corresponds to an Mr of 186,968. The protein exhibits 71% and 76% homology to skeletal and cardiac alpha 1 subunits, respectively. When compared with skeletal and cardiac alpha 1 isoforms, the rat brain protein is intermediate in size at the amino terminus and shorter at the carboxyl terminus. Multiple subtypes of this alpha 1 isoform cDNA were characterized. These are indicative of alternative splicing of a primary transcript and encode three variants between motif I and motif II and two within the S3 region of motif IV. Thus, multiple isoforms of this rat brain alpha 1 subunit are possible.