MilliporeSigma
  • Estimated Glomerular Filtration Rate; Laboratory Implementation and Current Global Status.
Estimated Glomerular Filtration Rate; Laboratory Implementation and Current Global Status.
Advances in chronic kidney disease (2018-03-04)
W Greg Miller, Graham R D Jones
ABSTRACT
In 2002, the Kidney Disease Outcomes Quality Initiative guidelines for identifying and treating CKD recommended that clinical laboratories report estimated glomerular filtration rate (eGFR) with every creatinine result to assist clinical practitioners to identify people with early-stage CKD. At that time, the original Modification of Diet in Renal Disease (MDRD) Study equation based on serum creatinine measurements was recommended for calculating eGFR. Because the MDRD Study equation was developed using a nonstandardized creatinine method, a Laboratory Working Group of the National Kidney Disease Education program was formed and implemented standardized calibration traceability for all creatinine methods from global manufacturers by approximately 2010. A modified MDRD Study equation for use with standardized creatinine was developed. The Chronic Kidney Disease Epidemiology Collaboration developed a new equation in 2009 that was more accurate than the MDRD Study equation at values above 60 mL/min/1.73 m2. As of 2017, reporting eGFR with creatinine is almost universal in many countries. A reference system for cystatin C became available in 2010, and manufacturers are in the process to standardize cystatin C assays. Equations for eGFR based on standardized cystatin C alone and with creatinine are now available from the Chronic Kidney Disease Epidemiology Collaboration and other groups.