- Differentiating PC12 cells to evaluate neurite densities through live-cell imaging.
Differentiating PC12 cells to evaluate neurite densities through live-cell imaging.
STAR protocols (2023-01-06)
Jordyn Karliner, Diane E Merry
PMID36602900
ABSTRACT
Although PC12 cells are a valuable tool in neuroscience research, previously published PC12 cell differentiation techniques fail to consider the variability in differentiation rates between different PC12 cell strains and clonal variants. Here, we present a comprehensive protocol to differentiate PC12 cells into equivalent neurite densities through live-cell imaging for morphological, immunocytochemical, and biochemical analyses. We detail steps on optimized substrate coating, plating techniques, culture media, validation steps, and quantification techniques.
Sigma-Aldrich
Bovine Serum Albumin solution, 7.5% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethylsulfonyl Fluoride, Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.