MilliporeSigma
  • Insulin-induced palmitoylation regulates the Cardiac Na+/Ca2+ exchanger NCX1.
Insulin-induced palmitoylation regulates the Cardiac Na+/Ca2+ exchanger NCX1.
Cell calcium (2022-03-02)
Caglar Gök, Alan D Robertson, William Fuller
ABSTRACT
The cardiac Na+/Ca2+ Exchanger (NCX1) controls Ca2+ extrusion from the cytosol by mediating bidirectional exchange of Na+ for Ca2+, and therefore controls cardiac relaxation. Insulin regulates Ca2+ handling in cardiac tissue through NCX1, however how insulin changes NCX1 activity is poorly understood. Palmitoylation is the only post-translational modification identified to alter NCX1 activity. Here we show that insulin triggers local structural re-arrangements within existing NCX1 dimers by inducing their palmitoylation, thus tunes NCX1 inactivation through a zDHHC5-dependent mechanism in multiple cell types. By activating fatty acid and fatty acyl CoA synthesis insulin promotes palmitoylation of the zDHHC5 active site, which leads to enhanced NCX1 palmitoylation. Our findings represent a new mechanism to regulate the palmitoylation of numerous zDHHC5 substrates.
Sigma-Aldrich
Anti-ZDHHC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triacsin C from Streptomyces sp.