MilliporeSigma
  • Myocardial Protecting Role of Glutamine in Patients with Low Ejection Fraction Undergoing Elective On-Pump Coronary Artery Bypass Graft Surgery.
Myocardial Protecting Role of Glutamine in Patients with Low Ejection Fraction Undergoing Elective On-Pump Coronary Artery Bypass Graft Surgery.
Vascular health and risk management (2022-04-13)
I Made Adi Parmana, Cindy Elfira Boom, Lisnawati Rachmadi, Dudy Arman Hanafy, Yunita Widyastuti, Muchtaruddin Mansyur, Bambang Budi Siswanto
ABSTRACT
Myocardial injury due to on-pump coronary artery bypass grafting (CABG) in patients with low ejection fraction (EF) is associated with poor outcomes. This study determines whether intravenous glutamine could protect the myocardium during on-pump CABG in patients with low EF. This was a double-blind, randomized controlled trial to assess glutamine as a myocardial protector during on-pump CABG in patients with left ventricle EF of 31-50%, conducted from January to October 2021. Patients in the glutamine group (n = 30) received 0.5 g/kg of 20% glutamine solution diluted with 0.9% NaCl up to 500 mL in total volume over a period of 24 hours. Patients in the control group (n = 30) received 0.9% NaCl over the same period. The primary outcomes were plasma troponin I and plasma glutamine levels. Secondary outcomes included α-ketoglutarate (α-KG) levels and histopathology scoring of the right atrial appendage tissue, plasma lactate levels, hemodynamic measurement, and morbidity. Twenty-nine patients from each group (58 in total) were included in the analysis. Plasma troponin I levels at 6 and 24 hours after cardiopulmonary bypass (CPB) were significantly lower in the glutamine than the control group (mean 3.43 ± 1.51 ng/mL vs mean 4.41 ± 1.89 ng/mL; p = 0.034; median 3.08 ng/mL [min-max: 1.30-6.59] vs median 3.77 ng/mL [min-max: 0.00-36.53]; p = 0.038, respectively). Plasma glutamine levels at 24 hours after CPB were significantly higher in the glutamine than the control group (mean 935.42 ± 319.10 μmol/L vs mean 634.79 ± 243.89 μmol/L, p = 0.001). Plasma lactate levels at 6 and 24 hours after CPB were significantly lower in the glutamine than the control group (median 5.30 mmol/L [min-max: 1.20-9.50] vs median 5.70 mmol/L [min-max: 2.80-11.30], p = 0.042; mean 2.08 ± 0.67 mmol/L vs mean 2.46 ± 0.69 mmol/L, p = 0.044, respectively). Myocardial injury score was significantly lower in the glutamine than the control group (mean 1.30 ± 0.24 vs mean 1.48 ± 0.26, p = 0.011). Perioperative administration of 0.5 g/kg intravenous glutamine solution over the period of 24 hours has myocardial protection effect in patients with low EF who undergo elective on-pump CABG.
Sigma-Aldrich
Glutamine and Glutamate Determination Kit