MilliporeSigma
  • Cyclization Reaction-Based Turn-on Probe for Covalent Labeling of Target Proteins.
Cyclization Reaction-Based Turn-on Probe for Covalent Labeling of Target Proteins.
Cell chemical biology (2020-01-29)
Hiroyuki Kojima, Yuki Fujita, Ryosuke Takeuchi, Yuka Ikebe, Nami Ohashi, Keiko Yamamoto, Toshimasa Itoh
ABSTRACT
Fluorescent molecules have contributed to basic biological research but there are currently only a limited number of probes available for the detection of non-enzymatic proteins. Here, we report turn-on fluorescent probes mediated by conjugate addition and cyclization (TCC probes). These probes react with multiple amino acids and exhibit a 36-fold greater emission intensity after reaction. We analyzed the reactions between TCC probes and nuclear receptors by electrospray ionization mass spectrometry, X-ray crystallography, spectrofluorometry, and fluorescence microscopy. In vitro analysis showed that probes consisting of a protein ligand and TCC could label vitamin D receptor and peroxisome proliferator-activated receptor γ. Moreover, we demonstrated that not only a ligand unit but also a peptide unit can label the target protein in a complex mixture.
Sigma-Aldrich
Rosetta 2(DE3) Competent Cells - Novagen, Novagen′s Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.