MilliporeSigma
  • The sustainable synthesis of peptidomimetics via chemoenzymatic tandem oxidation-Ugi reaction.
The sustainable synthesis of peptidomimetics via chemoenzymatic tandem oxidation-Ugi reaction.
RSC advances (2018-08-08)
Arleta Madej, Dominik Koszelewski, Daniel Paprocki, Anna Brodzka, Ryszard Ostaszewski
ABSTRACT
A simply and green synthetic protocol based on the selective laccase-oxidation of alcohol to a corresponding aldehyde and a following Ugi reaction in a micellar system made of SDS was developed and is reported herein. Special emphasis was placed on the metal-free chemoenzymatic tandem reaction based on alcohol oxidation strategies using molecular oxygen from air, followed by an Ugi reaction. The reaction was carried out without the use of a transition metal or organic solvents as a reaction medium. The presented protocol offers an efficient and environmentally friendly procedure.
Sigma-Aldrich
Laccase from Trametes versicolor, powder, light brown, ≥0.5 U/mg