MilliporeSigma
  • Improved solid-phase extraction method for systematic toxicological analysis in biological fluids.
Improved solid-phase extraction method for systematic toxicological analysis in biological fluids.
Journal of analytical toxicology (2001-04-13)
T Soriano, C Jurado, M Menéndez, M Repetto
ABSTRACT
A method for the simultaneous qualitative and quantitative determination of drugs of abuse (opiates, cocaine, or amphetamines) and prescribed drugs (tricyclic antidepressants, phenotiazines, benzodiazepines, etc.) in biological fluids--blood, urine, bile, and gastric contents--was developed. This procedure involves solid-phase extraction with Bond-Elut Certify columns followed by analysis by gas chromatography-nitrogen-phosphorus detection (GC-NPD) and confirmation by gas chromatography-mass spectrometry (GC-MS), after derivatization, when necessary. Pretreatment was performed on all samples: sonication for 15 min plus enzymatic hydrolysis with beta-glucuronidase in urine. With respect to the internal standards, nalorphine and trihexylamine were used for basic substances, allobarbital for acidic drugs, and prazepam for benzodiazepines. Acidic and basic compounds were extracted from different aliquots of samples at different pH levels: 6-6.5 for the acidic and neutral and 8-8.5 for the basic and the benzodiazepines. Several areas of experimental design were considered in the process of method optimization. These included internal standards, pH, sonication, flow rate and washing solvents. It was found that systematic analysis could be reliably performed using optimized extraction conditions. The recovery rates for the compounds tested were always higher than 61.02%.
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid