MilliporeSigma
  • Daldiniaeschsone A, a Rare Tricyclic Polyketide Having a Chromone Unit Fused to a δ-Lactone and Its Symmetrical Biphenyl Dimer, Daldiniaeschsone B, from an Endophytic Fungus Daldinia eschscholtzii SDBR-CMUNKC745.
Daldiniaeschsone A, a Rare Tricyclic Polyketide Having a Chromone Unit Fused to a δ-Lactone and Its Symmetrical Biphenyl Dimer, Daldiniaeschsone B, from an Endophytic Fungus Daldinia eschscholtzii SDBR-CMUNKC745.
Journal of fungi (Basel, Switzerland) (2021-06-03)
Natnicha Wutthiwong, Virayu Suthiphasilp, Aknarin Pintatum, Nakarin Suwannarach, Jaturong Kumla, Saisamorn Lumyong, Tharakorn Maneerat, Rawiwan Charoensup, Sarot Cheenpracha, Thunwadee Limtharakul, Stephen G Pyne, Surat Laphookhieo
ABSTRACT
Daldiniaeschsone A (1), a rare tricyclic polyketide having a chromone unit fused to a δ-lactone and its symmetrical 6,6'-biphenyl dimer, daldiniaeschsone B (2), together with three known compounds (3-5), were isolated from a plant-derived endophytic fungus, Daldinia eschscholtzii SDBR-CMUNKC745. Their structures were elucidated by extensive 1D and 2D NMR spectroscopic data and HRESIMS. All compounds showed α-glucosidase inhibitory activity with IC50 values ranging from 0.16-0.85 mM and compound 1 was the best α-glucosidase inhibitory activity (IC50 = 0.16 mM).
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)