MilliporeSigma
  • Mycobacterium tuberculosis up-regulates matrix metalloproteinase-1 secretion from human airway epithelial cells via a p38 MAPK switch.
Mycobacterium tuberculosis up-regulates matrix metalloproteinase-1 secretion from human airway epithelial cells via a p38 MAPK switch.
Journal of immunology (Baltimore, Md. : 1950) (2005-10-08)
Paul T G Elkington, Jenny E Emerson, Laura D C Lopez-Pascua, Cecilia M O'Kane, Donna E Horncastle, Joseph J Boyle, Jon S Friedland
ABSTRACT
Pulmonary cavitation is vital to the persistence and spread of Mycobacterium tuberculosis (MTb), but mechanisms underlying this lung destruction are poorly understood. Fibrillar type I collagen provides the lung's tensile strength, and only matrix metalloproteinases (MMPs) can degrade it at neutral pH. We investigated MTb-infected lung tissue and found that airway epithelial cells adjacent to tuberculosis (Tb) granulomas expressed a high level of MMP-1 (interstitial collagenase). Conditioned media from MTb-infected monocytes (CoMTb) up-regulated epithelial cell MMP-1 promoter activity, gene expression, and secretion, whereas direct MTb infection did not. CoMTb concurrently suppressed tissue inhibitor of metalloprotease-1 (TIMP-1) secretion, further promoting matrix degradation, and in Tb patients very low TIMP-1 expression was detected. MMP-1 up-regulation required synergy between TNF-alpha and G protein-coupled receptor signaling pathways. CoMTb stimulated p38 MAPK phosphorylation, and this is the point of TNF-alpha synergy with G protein-coupled receptor activation. Furthermore, p38 phosphorylation was the switch up-regulating MMP-1 activity and decreasing TIMP-1 secretion. Activated p38 localized to MMP-1-secreting airway epithelial cells in Tb patients. These data reveal a monocyte-epithelial cell network whereby MTb may drive tissue destruction, and they demonstrate that p38 phosphorylation is a key regulatory point in the generation of a matrix-degrading phenotype.