MilliporeSigma
  • Mice expressing the variant rs1143679 allele of ITGAM (CD11b) show impaired DC-mediated T cell proliferation.
Mice expressing the variant rs1143679 allele of ITGAM (CD11b) show impaired DC-mediated T cell proliferation.
Mammalian genome : official journal of the International Mammalian Genome Society (2019-11-02)
Justin T Avery, Rachel V Jimenez, Joseph L Blake, Tyler T Wright, Beatriz Leόn-Ruiz, Trenton R Schoeb, Alexander J Szalai, Daniel C Bullard
ABSTRACT
Genome-wide association studies (GWAS) and functional genomic analyses have implicated several ITGAM (CD11b) single-nucleotide polymorphisms (SNPs) in the development of SLE and other disorders. ITGAM encodes the αM chain of the β2 integrin Mac-1, a receptor that plays important roles in myeloid cell functions. The ITGAM SNP rs1143679, which results in an arginine to histidine change at amino acid position 77 of the CD11b protein, has been shown to reduce binding to several ligands and to alter Mac-1-mediated cellular response in vitro. Importantly, however, the potential contribution of this SNP variant to the initiation and/or progression of immune and inflammatory processes in vivo remains unexplored. Herein, we describe for the first time the generation and characterization of a mouse line expressing the 77His variant of CD11b. Surprisingly, we found that 77His did not significantly affect Mac-1-mediated leukocyte migration and activation as assessed using thioglycollate-induced peritonitis and LPS/TNF-α-induced dermal inflammation models. In contrast, expression of this variant did alter T cell immunity, as evidenced by significantly reduced proliferation of ovalbumin (OVA)-specific transgenic T cells in 77His mice immunized with OVA. Reduced antigen-specific T cell proliferation was also observed when either 77His splenic dendritic cells (DCs) or bone marrow-derived DCs were used as antigen-presenting cells (APCs). Although more work is necessary to determine how this alteration might influence the development of SLE or other diseases, these in vivo findings suggest that the 77His variant of CD11b can compromise the ability of DCs to induce antigen-driven T cell proliferation.
Product Description
Millipore
Thioglycollate medium, Microbiologically tested, NutriSelect® Plus