MilliporeSigma
  • Ammonium and glutamate released by neurons are signals regulating the nutritive function of a glial cell.
Ammonium and glutamate released by neurons are signals regulating the nutritive function of a glial cell.
The Journal of neuroscience : the official journal of the Society for Neuroscience (1997-04-01)
M Tsacopoulos, C L Poitry-Yamate, S Poitry
ABSTRACT
Glial cells transform glucose to a fuel substrate taken up and used by neurons. In the honeybee retina, photoreceptor neurons consume both alanine supplied by glial cells and exogenous proline. Ammonium (NH4+) and glutamate, produced and released in a stimulus-dependent manner by photoreceptor neurons, contribute to the biosynthesis of alanine in glia. Here we report that NH4+ and glutamate are transported into glia and that a transient rise in the intraglial concentration of NH4+ or of glutamate causes a net increase in the level of reduced nicotinamide adenine dinucleotides [NAD(P)H]. Biochemical measurements indicate that this is attributable to activation of glycolysis in glial cells by the direct action of NH4+ and glutamate on at least two enzymatic reactions: those catalyzed by phosphofructokinase (PFK; ATP:D-fructose-6-phosphotransferase, EC2.7.1.11) and glutamate dehydrogenase (GDH; L-glutamate:NAD oxidoreductase, deaminating; EC1.4.1.3). This activation leads to an increase in the production and release of alanine by glia. This signaling, which depends on the rate of conversion of NH4+ and glutamate to alanine and alpha-ketoglutarate, respectively, in the glial cells, raises the novel possibility of a tight regulation of the nutritive function of glia.
Product Description
Millipore
Pronase® Protease, Streptomyces griseus