MilliporeSigma
  • The cell-surface 5'-nucleotidase CD73 defines a functional T memory cell subset that declines with age.
The cell-surface 5'-nucleotidase CD73 defines a functional T memory cell subset that declines with age.
Cell reports (2021-11-11)
Fengqin Fang, Wenqiang Cao, Weikang Zhu, Nora Lam, Lingjie Li, Sadhana Gaddam, Yong Wang, Chulwoo Kim, Simon Lambert, Huimin Zhang, Bin Hu, Donna L Farber, Cornelia M Weyand, Jörg J Goronzy
ABSTRACT
Memory T cells exhibit considerable diversity that determines their ability to be protective. Here, we examine whether changes in T cell heterogeneity contribute to the age-associated failure of immune memory. By screening for age-dependent T cell-surface markers, we identify CD4 and CD8 memory T cell subsets that are unrelated to previously defined subsets of central and effector memory cells. Memory T cells expressing the ecto-5'-nucleotidase CD73 constitute a functionally distinct subset of memory T cells that declines with age. They resemble long-lived, polyfunctional memory cells but are also poised to display effector functions and to develop into cells resembling tissue-resident memory T cells (TRMs). Upstream regulators of differential chromatin accessibility and transcriptomes include transcription factors that facilitate CD73 expression and regulate TRM differentiation. CD73 is not just a surrogate marker of these regulatory networks but is directly involved in T cell survival.
Sigma-Aldrich
Collagenase type I, The collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means.