MilliporeSigma
  • Cytotoxicity of Ficus Crocata Extract on Cervical Cancer Cells and Protective Effect against Hydrogen Peroxide-Induced Oxidative Stress in HaCaT Non-Tumor Cells.
Cytotoxicity of Ficus Crocata Extract on Cervical Cancer Cells and Protective Effect against Hydrogen Peroxide-Induced Oxidative Stress in HaCaT Non-Tumor Cells.
Plants (Basel, Switzerland) (2021-01-23)
Brenda De la Cruz-Concepción, Mónica Espinoza-Rojo, Patricia Álvarez-Fitz, Berenice Illades-Aguiar, Macdiel Acevedo-Quiroz, Ana E Zacapala-Gómez, Napoleón Navarro-Tito, Hilda Jiménez-Wences, Francisco I Torres-Rojas, Miguel A Mendoza-Catalán
ABSTRACT
Oxidative stress causes several chronic diseases including cancer. Some chemotherapeutic agents are not selective against tumor cells, causing oxidative stress in non-tumor cells. This study aimed to evaluate the cytotoxic effect of acetone extract of Ficus crocata (Miq.) Mart. ex Miq. (F. crocata) leaves (Ace-EFc) on cervical cancer cells, as well as its protective effect on hydrogen peroxide (H2O2)-induced lipoperoxidation and cytotoxicity in non-tumor HaCaT cells. Antioxidant activity was determined using the DPPH and ABTS radicals. Cell viability and lipoperoxidation were determined with MTT and 1-methyl-2-phenylindole assays, respectively. A model of H2O2-induced cytotoxicity and oxidative damage in HaCaT cells was established. HaCaT cells were exposed to the extract before or after exposure to H2O2, and oxidative damage and cell viability were evaluated. Ace-EFc inhibited the DPPH and ABTS radicals and showed a cytotoxic effect on SiHa and HeLa cells. Furthermore, the extract treatment had a protective effect on hydrogen peroxide-induced lipoperoxidation and cytotoxicity, avoiding the increase in MalonDiAldehyde (MDA) levels and the decrease in cell viability (p < 0.001). These results suggest that the metabolites of F. crocata leaves possess antioxidant and cytoprotective activity against oxidative damage. Thus, they could be useful for protecting cells from conditions that cause oxidative stress.
Sigma-Aldrich
MTT Cell Growth Assay Kit, MTT Cell Growth Assay is a colorimetric assay that can be used for either proliferation or complement-mediated cytotoxicity assays.