MilliporeSigma
  • SARS-CoV-2 3a expression, purification, and reconstitution into lipid nanodiscs.
SARS-CoV-2 3a expression, purification, and reconstitution into lipid nanodiscs.
Methods in enzymology (2021-06-09)
David M Kern, Stephen G Brohawn
ABSTRACT
The SARS-CoV-2 3a protein is a putative ion channel implicated in virus life cycle and pathogenesis. We recently expressed, purified, and reconstituted 3a into lipid nanodiscs to solve its structure by cryo-EM to 2.1Å resolution. In this chapter, we describe methods we developed in order to facilitate the study of this protein in other laboratories. We emphasize factors that enabled rapid progression from gene sequence to reconstituted protein (3 weeks in the case of 3a) and provide general observations and tips for adapting these protocols to other membrane proteins of interest.
Product Description
Millipore
Benzonase® Nuclease, Purity > 90%