MilliporeSigma
[Squamous papillomas of the esophagus].
Minerva chirurgica (1992-08-01)
C Bazzoni, M Ongari, M Alleva, A Crespi, C Sguazzini, C Lombardi
ABSTRACT
The paper reports 14 cases of squamous papilloma of the esophagus which were removed using an endoscopic method: this is a comparatively rare benign pathology of which an increasing number of cases have been recently observed. Endoscopic controls were carried out in all cases and the results of the follow-up are reported.