MilliporeSigma
  • Protective effects of amentoflavone on Lamin A-dependent UVB-induced nuclear aberration in normal human fibroblasts.
Protective effects of amentoflavone on Lamin A-dependent UVB-induced nuclear aberration in normal human fibroblasts.
Bioorganic & medicinal chemistry letters (2011-09-16)
Nok-Hyun Park, Chan-Woo Lee, Ji-Hyun Bae, Yong Joo Na
ABSTRACT
Lamin A-dependent nuclear aberration and DNA damage was found in premature aging disease or normally old individuals. In this study, UVB irradiation was used as a cellular senescence inducer, and it was found that Lamin A and phospho-H2AX protein was increased by UVB treatment on normal human fibroblast. Lamin A-dependent morphological nuclear defect was observed in UVB treated fibroblast. Amentoflavone, a well known biflavonoid, inhibited the increase of Lamin A or phospho-H2AX protein in dose dependent manner which was induced by UVB irradiation, and also protected nuclear aberration dramatically. These results indicated that amentoflavone is an anti-aging candidate for UVB related skin aging process.
Sigma-Aldrich
Amentoflavone, ≥98.0% (HPLC)
Supelco
Amentoflavone, analytical standard