MilliporeSigma
  • Proteomics and metabolomics of raw rhubarb and wine-processed rhubarb in the treatment of rats with intracerebral hemorrhage.
Proteomics and metabolomics of raw rhubarb and wine-processed rhubarb in the treatment of rats with intracerebral hemorrhage.
Annals of translational medicine (2021-01-26)
Kun Wang, Dongsheng Guan, Xu Zhao, Dongge Qiao, Yanhua Yang, Yinglin Cui
ABSTRACT
To explore the mechanisms of raw rhubarb and wine-processed rhubarb treatment in a rat model of intracerebral hemorrhage (ICH). After adapting to their environment, 30 male Wistar rats were divided into 5 treatment groups: blank control group (CK) (normal saline), sham operation group (SICH) (normal saline), ICH model group (ICH) (normal saline), ICH + raw rhubarb enema group (RO) (raw rhubarb 3.60 g/kg), and ICH + wine-processed rhubarb enema group (WRO) (wine-processed rhubarb 3.60 g/kg). The rhubarb enema (once a day) was given since 3 days before ICH treatment, and was consistently given until 12 hours, 24 hours, 3 days, and 7 days post operation. Serum oxidative stress and inflammatory markers were detected by ELISA, and differences between raw rhubarb and wine-processed rhubarb treatment in ICH rats were screened by proteomics and metabolomics. Both rhubarb treatments effectively improved oxidative stress and inflammatory responses. After ICH, the proteins and metabolites in the brain tissue were significantly altered. Compared with raw rhubarb, wine-processed rhubarb had a better protective effect by dysregulating amino acid metabolism in the ICH model. Our study provides a basis for further research into the mechanisms of rhubarb treatment from different processing technologies, and promotes the application of rhubarb as a therapeutic approach to ICH.