- A Stable Biotin-Streptavidin Surface Enables Multiplex, Label-Free Protein Detection by Aptamer and Aptamer-Protein Arrays Using Arrayed Imaging Reflectometry.
A Stable Biotin-Streptavidin Surface Enables Multiplex, Label-Free Protein Detection by Aptamer and Aptamer-Protein Arrays Using Arrayed Imaging Reflectometry.
While label-free multiplex sensor technology enables "mixing and matching" of different capture molecules in principle, in practice this has been rarely (if ever) demonstrated. To fill this gap, we developed protocols for the preparation of mixed aptamer-protein arrays on the arrayed imaging reflectometry (AIR) sensing platform using streptavidin as a common attachment point for both biotinylated proteins and aptamers. Doing so required overcoming the noted instability of dried streptavidin monolayers on surfaces. After characterizing this degradation, stable surfaces were obtained using a commercial microarray product. Microarraying through the layer of stabilizer then provided mixed aptamer-antibody arrays. We demonstrate that sensor arrays prepared in this manner are suitable for several probes (thrombin and TGF-β1 aptamers; avi-tagged protein) and targets.