MilliporeSigma
  • A sensitive immunoassay based on fluorescence resonance energy transfer from up-converting nanoparticles and graphene oxide for one-step detection of imidacloprid.
A sensitive immunoassay based on fluorescence resonance energy transfer from up-converting nanoparticles and graphene oxide for one-step detection of imidacloprid.
Food chemistry (2020-08-03)
Yirong Guo, Rubing Zou, Fangfang Si, Wenlong Liang, Tianyi Zhang, Yunyun Chang, Xvsheng Qiao, Jinhao Zhao
ABSTRACT
In this study, a fluorescence resonance energy transfer (FRET) immunoassay based on graphene oxide (GO) and up-converting nanoparticles (UCNPs) was established for rapid detection of imidacloprid, a commonly-used insecticide. Under 980 nm near-infrared light excitation, emission of UCNPs at 542 nm can be absorbed by the energy acceptor GO. The carboxyl-functionalized GO and UCNPs were coupled with competitive antigen and antibody against imidacloprid. After optimization, the FRET immunoassay showed a wide detection range of 0.08-50 ng/mL to imidacloprid, with cross-reaction toward other three neonicotinoids including imidaclothiz (74.4%), thiacloprid (36.9%) and clothianidin (31.9%). The average recoveries of spiked water, Chinese cabbage, cucumber, honey and tea samples were 76.8%-101.8%. The accuracy and reliability of the FRET immunoassay were verified by UPLC-MS/MS with a good correlation (R2 = 0.9816). In a summary, this study provides a sensitive and one-step method for monitoring imidacloprid residue in food and environmental samples within 1 h.
Sigma-Aldrich
Sodium chloroacetate, 98%