MilliporeSigma
  • Effects of Acute Eccentric Exercise on Appetite-Related Hormones and Food Preferences in Men.
Effects of Acute Eccentric Exercise on Appetite-Related Hormones and Food Preferences in Men.
American journal of men's health (2019-07-03)
Shaea Alkahtani, Abdulaziz Aldayel, Mark Hopkins
ABSTRACT
Eccentric exercise has been suggested to improve muscle atrophy, muscle function, and insulin sensitivity. The aim of this study was to examine the effect of acute eccentric exercise on appetite-related hormones, food preferences, and food intake. Fourteen moderately active men were recruited to participate in this study (age 24.2 ± 5.5 years; BMI 23.4 ± 3.3 kg/m2; VO2max 48.9 ± 3.1 ml/kg/min). Three different conditions were implemented; no exercise, flat running "inclination 0" and downhill running "inclination -12%." Appetite-related hormones, subjective appetite sensations, food preference and reward, and ad libitum food intake were measured at pre-, immediately post-, and 24 h post exercise. There were no significant median changes in total ghrelin or pancreatic peptide concentrations between conditions. There were also no median differences in subjective appetite ratings or energy intake between conditions, but the median change in explicit liking of sweet versus savory foods differed significantly between pre-exercise and 24 h post exercise (p = .013). Post-hoc analysis observed a significant difference in the pre-exercise to 24 h post exercise change between front running and downhill running (p = .023), and indicated greater liking of savory foods over sweet foods in downhill running than front running. However, no further differences were seen between conditions for the remaining food preference parameters, suggesting there were no systematic trends in these data. In conclusion, there was no effect of front and downhill running on eating behavior as compared to a nonexercise control condition, but these data need to be replicated in a larger and more heterogeneous sample.