MilliporeSigma
  • Neuroprotective effect of intravitreal triamcinolone acetonide against photoreceptor apoptosis in a rabbit model of subretinal hemorrhage.
Neuroprotective effect of intravitreal triamcinolone acetonide against photoreceptor apoptosis in a rabbit model of subretinal hemorrhage.
Investigative ophthalmology & visual science (2008-04-19)
Robert B Bhisitkul, Bryan J Winn, On-Tat Lee, Joshua Wong, Daniel de Souza Pereira, Travis C Porco, Xining He, Paul Hahn, Joshua L Dunaief
ABSTRACT
To study photoreceptor apoptosis and iron migration as mechanisms of retinotoxicity in a rabbit model of subretinal hemorrhage (SRH) and to assess intravitreal triamcinolone acetonide (IVTA) for anti-apoptotic and neuroprotective effects. In adult rabbits, eyes were studied histologically after subretinal injection of autologous blood. For comparisons of control eyes with eyes injected with 2 mg IVTA, morphometric analysis was performed with light microscopy, whereas apoptosis was quantified with terminal dUTP nick end labeling (TUNEL) and fluorescence microscopy. Localization of retinal iron was assessed with Perls' stain. Photoreceptor degeneration was initiated 48 hours after exposure to subretinal blood and progressed over 7 days. Increased TUNEL positivity demonstrating apoptotic cell death was associated with SRH and photoreceptor loss. VIP-Perls staining demonstrated iron in the photoreceptor layer and retinal pigment epithelium that correlated with photoreceptor degeneration. Treatment with IVTA enhanced photoreceptor cell survival by 11% at 48 hours and by 45% at 72 hours (P = 0.01) and reduced photoreceptor apoptosis ratios by 25% at 48 hours (P = 0.006). Photoreceptor toxicity caused by SRH occurs at least in part by apoptosis and is associated with iron migration to the photoreceptor layer. Treatment with IVTA reduced photoreceptor loss and apoptosis, indicating a neuroprotective action. Therapies to target SRH may augment anti-VEGF treatments in exudative age-related macular degeneration and other diseases of choroidal neovascularization.
Sigma-Aldrich
FragEL DNA Fragmentation Detection Kit, Fluorescent - TdT Enzyme