MilliporeSigma
  • An endophytic microbe from an unusual volcanic swamp corn seeks and inhabits root hair cells to extract rock phosphate.
An endophytic microbe from an unusual volcanic swamp corn seeks and inhabits root hair cells to extract rock phosphate.
Scientific reports (2017-10-19)
Hanan R Shehata, Christopher Dumigan, Sophia Watts, Manish N Raizada
ABSTRACT
In the animal microbiome, localization of microbes to specific cell types is well established, but there are few such examples within the plant microbiome which includes endophytes. Endophytes are non-pathogenic microbes that inhabit plants. Root hairs are single cells, equivalent to the nutrient-absorbing intestinal microvilli of animals, used by plants to increase the root surface area for nutrient extraction from soil including phosphorus (P). There has been significant interest in the microbiome of intestinal microvilli but less is known about the root hair microbiome. Here we describe a bacterial endophyte (3F11) from Zea nicaraguensis, a wild corn discovered in a Nicaraguan swamp above rock-P lava flowing from the San Cristobal volcano. Rock-P is insoluble and a major challenge for plants. Following seed coating and germination on insoluble-P, the endophyte colonized epidermal surfaces, ultimately colonizing root hairs intracellularly. The endophyte promoted root hair growth and secreted acids to solubilize rock-P for uptake by a larger root hair surface. The most interesting observation was that a seed-coated endophyte targeted and colonized a critical cell type, root hair cells, consistent with earlier studies. The endophyte maintained its targeting ability in two evolutionary divergent hosts, suggesting that the host recognition machinery is conserved.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 70 purifications