MilliporeSigma
  • Preparation of magnetic zeolitic imidazolate framework-67 composites for the extraction of phthalate esters from environmental water samples.
Preparation of magnetic zeolitic imidazolate framework-67 composites for the extraction of phthalate esters from environmental water samples.
Analytical methods : advancing methods and applications (2020-10-01)
Qingsong Zhang, Guangyang Liu, Xiaolin Cao, Jungang Yin, Ziping Zhang
ABSTRACT
In this paper, magnetic ZIF-67 composites were prepared by an in situ method and utilized in the magnetic solid-phase extraction of phthalate esters. The obtained magnetic ZIF-67 composites were characterized in detail, and the adsorption performance was evaluated. The results showed that ZIF-67 crystals in the composites had a chamfered cubic shape with about 2.0 μm diameter and Fe3O4 was attached on the surface of ZIF-67. The parameters affecting magnetic solid-phase extraction efficiency were optimized including the adsorption and desorption conditions. Under the optimum conditions, magnetic solid-phase extraction coupled with gas chromatography-mass spectrometry for analyzing seven phthalate esters in environmental water samples was established. All the analytes exhibited good linearity (1-200 ng mL-1) with correlation coefficients higher than 0.9950. The relative standard deviations were between 4.3% and 8.9%. The developed method showed high sensitivity and had good potential for analyzing trace phthalate esters in water samples.
Product Description
Supelco
Dihexyl phthalate, analytical standard