MilliporeSigma
  • Secretory expression in Bacillus subtilis and biochemical characterization of a highly thermostable polyethylene terephthalate hydrolase from bacterium HR29.
Secretory expression in Bacillus subtilis and biochemical characterization of a highly thermostable polyethylene terephthalate hydrolase from bacterium HR29.
Enzyme and microbial technology (2020-12-31)
Xingxiang Xi, Kefeng Ni, Helong Hao, Yuepeng Shang, Bo Zhao, Zhen Qian
ABSTRACT
The environmental threat posed by disposal of plastic wastes has drawn extensive attention in recent years wherein polyethylene terephthalate (PET) constitutes one of the major plastic materials in the wastes. Recycling of PET wastes into reusable materials effectively overcomes its accumulation in the environment and can be achieved by mechanical, chemical, and biological processes. In comparison to the other methods, enzymatic treatment utilizing PET hydrolyzing enzymes (PETases) is environmental-friendly which avoids the use of hazardous chemicals. In this study, we report on the secretory expression in Bacillus subtilis a PETase (BhrPETase) from the bacterium HR29, a close homologue of the leaf-branch compost cutinase (LCC) with 94 % sequence identity. The expression titer of BhrPETase reached 0.66 g/L in an engineered chaperone-overexpression Bacillus subtilis strain, and the biochemical characterization of BhrPETase for the first time revealed its high hydrolyzing activity towards amorphous PET in comparison to two reported PET hydrolyzing enzymes LCC and IsPETase, which were expressed under the same expression conditions in Bacillus subtilis in our study. Most intriguingly, purified BhrPETase displayed a melting temperature as high as 101 °C. To our knowledge it is the most thermostable bacterial PETase characterized so far. The superior activity and thermostability of BhrPETase rendered it one of the most promising PETases for plastic waste recycling and bioremediation applications in the future.
Sigma-Aldrich
4-Nitrophenyl octanoate, ≥90.0% (GC)