MilliporeSigma
  • Circulating Levels of Visceral Adipose Tissue-Derived Serine Protease Inhibitor (Vaspin) Appear as a Marker of Musculoskeletal Pain Disability.
Circulating Levels of Visceral Adipose Tissue-Derived Serine Protease Inhibitor (Vaspin) Appear as a Marker of Musculoskeletal Pain Disability.
Diagnostics (Basel, Switzerland) (2020-10-15)
Nader Tarabeih, Alexander Kalinkovich, Adel Shalata, Gregory Livshits
ABSTRACT
Musculoskeletal pain (MSP), specifically low back pain (LBP), is often associated with several adipose tissue-derived cytokines (adipokines) and body composition, but their correlations with the LBP-related disability/severity phenotypes remain poorly understood. In this cross-sectional study, two self-reported validated questionnaires were used to collect back pain and disability data in an ethnically homogeneous family-based population sample (N = 1078). Plasma levels of relatively new adipokines, vaspin and adipsin, were detected by ELISA. Body composition parameters, including fat, skeletal muscle mass, extracellular water (ECW), and others were assessed through bioelectrical impedance analysis (BIA) technology. Statistical analysis was conducted, accounting for the familial composition of the sample. The multiple regression analyses with four LBP-related phenotypes as dependent variables consistently showed, for the first time, the significant associations with vaspin levels, regardless of other covariates. The odds ratios (OR)/SD ranged between 1.24 (95%CI = 1.03-1.50) and 1.33 (95%CI = 1.07-1.64), depending on the LBP phenotype. Among the tested body composition covariates, only ECW levels displayed consistent and highly significant associations with all tested LBP phenotypes (OR from 1.43, 95%CI = 1.14-1.79 to 1.68, 95%CI = 1.26-2.24). The results clearly suggest that circulating concentrations of vaspin and ECW levels could serve as biomarkers of MSP/LBP severity and complications.
Sigma-Aldrich
Human Adipsin / Factor D ELISA Kit, for serum, plasma, cell culture supernatant and urine