MilliporeSigma
  • Human CYP2D6 in the Brain Is Protective Against Harmine-Induced Neurotoxicity: Evidence from Humanized CYP2D6 Transgenic Mice.
Human CYP2D6 in the Brain Is Protective Against Harmine-Induced Neurotoxicity: Evidence from Humanized CYP2D6 Transgenic Mice.
Molecular neurobiology (2020-08-08)
Marlaina R Stocco, Cole Tolledo, Fariba Baghai Wadji, Frank J Gonzalez, Sharon Miksys, Rachel F Tyndale
ABSTRACT
CYP2D6 metabolically inactivates several neurotoxins, including beta-carbolines, which are implicated in neurodegenerative diseases. Variation in CYP2D6 within the brain may alter local inactivation of neurotoxic beta-carbolines, thereby influencing neurotoxicity. The beta-carboline harmine, which induces hypothermia and tremor, is metabolized by CYP2D6 to the non-hypothermic/non-tremorgenic harmol. Transgenic mice (TG), expressing human CYP2D6 in addition to their endogenous mouse CYP2D, experience less harmine-induced hypothermia and tremor compared with wild-type mice (WT). We first sought to elucidate the role of CYP2D in general within the brain in harmine-induced hypothermia and tremor severity. A 4-h intracerebroventricular (ICV) pretreatment with the CYP2D inhibitor propranolol increased harmine-induced hypothermia and tremor in TG and increased harmine-induced hypothermia in WT. We next sought to specifically demonstrate that human CYP2D6 expressed in TG brain altered harmine response severity. A 24-h ICV propranolol pretreatment, which selectively and irreversibly inhibits human CYP2D6 in TG brain, increased harmine-induced hypothermia. This 24-h pretreatment had no impact on harmine response in WT, as propranolol is not an irreversible inhibitor of mouse CYP2D in the brain, thus confirming no off-target effects of ICV propranolol pretreatment. Human CYP2D6 activity in TG brain was sufficient in vivo to mitigate harmine-induced neurotoxicity. These findings suggest that human CYP2D6 in the brain is protective against beta-carboline-induced neurotoxicity and that the extensive interindividual variability in CYP2D6 expression in human brain may contribute to variation in susceptibility to certain neurotoxin-associated neurodegenerative disorders.
Sigma-Aldrich
Harmine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Dextrorphan