MilliporeSigma
  • Influence of hypergastrinemia secondary to long-term proton pump inhibitor treatment on ECL cell tumorigenesis in human gastric mucosa.
Influence of hypergastrinemia secondary to long-term proton pump inhibitor treatment on ECL cell tumorigenesis in human gastric mucosa.
Pathology, research and practice (2020-08-28)
Atsushi Tatsuguchi, Shintaro Hoshino, Noriyuki Kawami, Katya Gudis, Tsutomu Nomura, Akira Shimizu, Katsuhiko Iwakiri
ABSTRACT
Proton pump inhibitor (PPI) therapy causes hypergastrinemia, which could promote the development and progression of neuroendocrine tumors (NETs). Concerns have been raised about the safety of long-term PPI use due to a possible increased risk of NETs. This study aimed to investigate the association between hypergastrinemia and the risk of NETs. Twenty outpatients presenting with serum gastrin levels greater than 400 pg/mL after long-term PPI treatment were registered in this study. Immunohistochemical analyses for chromogranin A (CgA), Ki67, gastrin and CCK/B gastrin receptor (CCKBR) were performed, and positive cell numbers were counted. There were no NET or gastric epithelial neoplasia cases observed among any of the 20 patients examined throughout the PPI treatment period. Histologically, ECL cell hyperplasia were shown in all patients. However, no relationship was found between serum gastrin levels and the number of CgA positive ECL cells. There was also no relationship between serum gastrin levels and the proportion of Ki67 positive cells or the density of CCKBR positive cells. The data indicate no relationship may exist between NETs and hypergastrinemia secondary to PPI treatment in patients having no, or mild, atrophic gastritis.