MilliporeSigma
  • RNA-binding protein Musashi2 regulates Hippo signaling via SAV1 and MOB1 in pancreatic cancer.
RNA-binding protein Musashi2 regulates Hippo signaling via SAV1 and MOB1 in pancreatic cancer.
Medical oncology (Northwood, London, England) (2020-08-12)
Haiyan Yang, Jiong Hu, Jingde Chen, Zhiqin Chen, Feng Jiao, Jiujie Cui, Ming Quan, Liwei Wang
ABSTRACT
Musashi 2 (MSI2), a member of the Musashi RNA-binding family, is reported to be an oncoprotein in pancreatic ductal adenocarcinoma (PDAC), but the mechanisms of MSI2 in the development and progression of PDAC have not been fully demonstrated. In this research, we studied the clinical significance, biologic effects and the underlying mechanism of MSI2 in the progression of PDAC. The expression of MSI2, Mps-binding protein 1 (MOB1) and Salvador family WW domain-containing protein 1 (SAV1) in PDAC tissues were analyzed immunohistochemically. The biologic effects of MSI2 regarding PDAC cell proliferation, migration and invasion were studied using gain- and loss-of-function assays. MSI2 regulated Hippo signaling pathway via SAV1 and MOB1 was tested in several PDAC cell lines, and the mechanisms were studied using molecular biologic methods. The expression of MSI2 was significantly increased in PDAC cell lines and tissues, and positively associated with tumor poorer differentiation, lymph nodes metastasis and TNM stages. Overexpression of MSI2 promoted PDAC cells proliferation, migration and invasion. Further studies demonstrated that MSI2 regulated the Hippo signaling pathway via directly binding to the mRNAs of SAV1 and MOB1, and controlled the translation and stability of SAV1 and the translation of MOB1. This study demonstrated that MSI2 regulated the Hippo signaling pathway via suppressing SAV1 and MOB1 at post-transcriptional level and promoted PDAC progression.
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%