MilliporeSigma
  • Tumor-Educated Platelets and Angiogenesis in Glioblastoma: Another Brick in the Wall for Novel Prognostic and Targetable Biomarkers, Changing the Vision from a Localized Tumor to a Systemic Pathology.
Tumor-Educated Platelets and Angiogenesis in Glioblastoma: Another Brick in the Wall for Novel Prognostic and Targetable Biomarkers, Changing the Vision from a Localized Tumor to a Systemic Pathology.
Cells (2020-01-30)
Rolando Campanella, Laura Guarnaccia, Chiara Cordiglieri, Elena Trombetta, Manuela Caroli, Giorgio Carrabba, Nicla La Verde, Paolo Rampini, Chiara Gaudino, Antonella Costa, Sabino Luzzi, Giovanna Mantovani, Marco Locatelli, Laura Riboni, Stefania Elena Navone, Giovanni Marfia
ABSTRACT
: Circulating platelets (PLTs) are able to affect glioblastoma (GBM) microenvironment by supplying oncopromoter and pro-angiogenic factors. Among these mediators, sphingosine-1-phophate (S1P) has emerged as a potent bioactive lipid enhancing cell proliferation and survival. Here, we investigated the effect of "tumor education", characterizing PLTs from GBM patients in terms of activation state, protein content, and pro-angiogenic potential. PLTs from healthy donors (HD-PLTs) and GBM patients (GBM-PLTs) were collected, activated, and analyzed by flow cytometry, immunofluorescence, and Western blotting. To assess the pro-angiogenic contribution of GBM-PLTs, a functional cord formation assay was performed on GBM endothelial cells (GECs) with PLT-releasate. GBM-PLTs expressed higher positivity for P-selectin compared to HD-PLTs, both in basal conditions and after stimulation with adenosine triphosphate (ADP) and thrombin receptor activating peptide (TRAP). PLTs showed higher expression of VEGFR-1, VEGFR-2, VWF, S1P, S1PR1, SphK1, and SPNS. Interestingly, increased concentrations of VEGF and its receptors VEGFR1 and VEGFR2, VWF, and S1P were found in GBM-PLT-releasate with respect to HD-PLTs. Finally, GBM-PLT-releasate showed a pro-angiogenic effect on GECs, increasing the vascular network's complexity. Overall, our results demonstrated the contribution of PLTs to GBM angiogenesis and aggressiveness, advancing the potential of an anti-PLT therapy and the usefulness of PLT cargo as predictive and monitoring biomarkers.