MilliporeSigma
KMT2A regulates cervical cancer cell growth through targeting VDAC1.
Aging (2020-05-22)
Changlin Zhang, Yijun Hua, Huijuan Qiu, Tianze Liu, Qian Long, Wei Liao, Jiehong Qiu, Nang Wang, Miao Chen, Dingbo Shi, Yue Yan, Chuanbo Xie, Wuguo Deng, Tian Li, Yizhuo Li
ABSTRACT
Cervical cancer is an aggressive cutaneous malignancy, illuminating the molecular mechanisms of tumorigenesis and discovering novel therapeutic targets are urgently needed. KMT2A is a transcriptional co-activator regulating gene expression during early development and hematopoiesis, but the role of KMT2A in cervical cancer remains unknown. Here, we demonstrated that KMT2A regulated cervical cancer growth via targeting VADC1. Knockdown of KMT2A significantly suppressed cell proliferation and migration and induced apoptosis in cervical cancer cells, accompanying with activation of PARP/caspase pathway and inhibition of VADC1. Overexpression of VDAC1 reversed the KMT2A knockdown-mediated regulation of cell proliferation, migration and apoptosis. The in vivo results from a cervical cancer xenograft mouse model also validated that KMT2A knockdown suppressed tumor growth by inhibiting VDAC1, whereas KMT2A overexpression promoted cervical cancer growth. Moreover, analyses of Biewenga cervix database and clinical samples showed that both KMT2A and VDAC1 were upregulated in cervix squamous cell carcinoma compared with cervix uteri tissues, and their expression was negatively correlated with the differentiation grade of cervical cancer. Our results therefore indicated that the KMT2A/VDAC1 signaling axis may be a potential new mechanism of cervical carcinogenesis.