MilliporeSigma
  • Altered expressions and splicing profiles of Acin1 transcripts differentially modulate brown adipogenesis through an alternative splicing mechanism.
Altered expressions and splicing profiles of Acin1 transcripts differentially modulate brown adipogenesis through an alternative splicing mechanism.
Biochimica et biophysica acta. Gene regulatory mechanisms (2020-07-07)
Ying-Chin Lin, Yi-Han Lu, Yuan-Chii Lee, Ching-Sheng Hung, Jung-Chun Lin
ABSTRACT
Apoptotic chromatin condensation inducer in the nucleus (also referred as Acin1) was first characterized as an RNA-binding protein involved in apoptosis. In later reports, Acin1 was identified as an auxiliary component of the exon junction complex (EJC) which is assembled throughout pre-messenger RNA splicing. In this study, results of whole-transcriptome analyses revealed reduced expressions and reprogrammed splicing profiles of Acin1 transcripts throughout development of brown adipose tissues (BATs) that execute non-shivering thermogenesis in small rodents and infants by consuming lipids. Depletion of endogenous Acin1 isoforms led to activation of brown adipogenic signatures in mouse C3H10T1/2 fibroblasts. Nevertheless, overexpressions of the Acin1-L or Acin1-S isoform exerted discriminative influences on brown adipogenesis and reprogramming of the expression of serine/arginine-rich splicing factor 3 (SRSF3) through an alternative splicing-coupled nonsense-mediated decay mechanism in a sequence-specific manner. Moreover, the Acin1-SRSF3 axis constitutes a regulatory pathway that participates in the brown adipocyte-related splicing network. Taken together, the interplay between accessory EJC components and splicing regulators constitutes an emerging mechanism for differentially manipulating the activity of brown adipogenesis via alternative splicing network.