- The impact of TP53BP1 and MLH1 on metastatic capability in cases of locally advanced prostate cancer and their usefulness in clinical practice.
The impact of TP53BP1 and MLH1 on metastatic capability in cases of locally advanced prostate cancer and their usefulness in clinical practice.
Lymph node (LN) metastases increase the risk of death from prostate cancer (CaP). The dysfunction of factors responsible for DNA injury detection may promote the evolution of localized primary tumors into the metastatic form. In this study, 52 cases of CaP were analyzed. The cases were divided into groups of CaP without metastases (N0), with metastases to the LNs (N+), and metastatic LN tissue. Immunohistochemical examinations were performed with antibodies against MDC1, TP53BP1, MLH1, MSH2, MSH6, and PMS2. Statistical analysis showed lower nuclear expression of TP53BP1 in N+ cases than in N0 cases (P = 0.026). Nuclear TP53BP1 expression was lower in LN cases than in N+ cases (P = 0.019). Statistical analysis showed lower nuclear expression of MLH1 in N+ cases than in to N0 cases (P = 0.003). Decreased expression of both MLH1 and TP53B1 were demonstrated in N+ cases of CaP. This observation could help to determine the risk of nodal metastasis, and to select appropriate treatment modalities for patients with locally advanced CaP.