MilliporeSigma
  • A Sphingosine-1-Phosphate Modulator Ameliorates Polycystic Kidney Disease in Han:SPRD Rats.
A Sphingosine-1-Phosphate Modulator Ameliorates Polycystic Kidney Disease in Han:SPRD Rats.
American journal of nephrology (2019-11-07)
Xin Li, Ming Wu, Limin Chen, Junyan Lu, Guo Li, Lili Fu, Na Qi, Ye Lin, Zhongya Sun, Xueqi Wang, Hao Zhang, Jingqiu Liu, Hualiang Jiang, Lin Li, Chang-Lin Mei, Cheng Luo
ABSTRACT
Inflammation plays an important role in polycystic kidney disease (PKD). Cordyceps sinensis, a prized -Chinese medicinal herb, exerts anti-tumor, anti-inflammatory and anti-metastatic effects and benefits patients with kidney diseases. The aim of this study was to test the efficacy of FTY720, an immunosuppressant derived from C. sinensis, in a rat cystic kidney disease model, and explore its underlining mechanism. Male wild type and Cy/+ Han:SPRD rats were treated with FTY720 at 3 and 10 mg/kg/day for 5 weeks and 12 weeks by gavage. Blood and kidney were collected for functional, morphological, RNA, and protein analysis. Inflammation is activated in Cy/+ Han:SPRD rats. Inflammatory cytokines including interleukin 6 and tumor necrosis factor alpha were upregulated and inflammation-related pathways were activated, such as nuclear factor κB and signal transducer and activator of transcription 3 (STAT3) pathways. Furthermore, the bioactive sphingolipid mediator sphingosine-1-phosphate (S1P), a regulator of inflammation, was accumulated in the Cy/+ Han:SPRD rats. FTY720 significantly reduced cyst growth and delayed disease progression by reducing the accumulation of S1P, thereby inhibiting inflammatory responses. FTY720 treatment reduced the expression of inflammatory cytokines and attenuated the activation of NK-κB and STAT3 pathways in Cy/+ Han:SPRD rats. It suggests that FTY720 may serve as a therapeutic agent for clinical autosomal dominant PKD treatment.