MilliporeSigma
  • Aberrant pregnancy-associated plasma protein-A expression in breast cancers prognosticates clinical outcomes.
Aberrant pregnancy-associated plasma protein-A expression in breast cancers prognosticates clinical outcomes.
Scientific reports (2020-08-15)
Prashanth Prithviraj, Matthew Anaka, Erik W Thompson, Revati Sharma, Marzena Walkiewicz, Candani S A Tutuka, Andreas Behren, George Kannourakis, Aparna Jayachandran
ABSTRACT
Elevated levels of pregnancy-associated plasma protein-A (PAPP-A) have been implicated in the pathogenesis of various malignancies, including breast cancers. Breast cancer is one of the most frequent carcinomas and is the second most common cancer type detected in women of child-bearing age. Throughout pregnancy PAPP-A is produced and secreted by the placental syncytiotrophoblast cells; co-incidentally pregnancy-associated breast cancers often have an aggressive clinical course. The components of the PAPP-A/IGF axis was assessed in a panel of breast cancer cell lines. Using neutralising antibodies the impact of PAPP-A/IGF axis on cell motility was evaluated. PAPP-A was expressed in four of the twelve breast cancer cell lines tested. Blocking PAPP-A and IGFBP4 with neutralising antibodies significantly decreased motiliy of MDA-MB-231 cells. Upregulation of PAPP-A expression in breast tumours resulted in a trend towards worse overall survival. Notably, PAPP-A expression also positively correlated with epithelial-to-mesenchymal transition markers. In conclusion, these results indicate that PAPP-A plays an important role in breast cancer progression and it may be a promising therapeutic target in breast cancer.