MilliporeSigma
  • LRP::FLAG Reduces Phosphorylated Tau Levels in Alzheimer's Disease Cell Culture Models.
LRP::FLAG Reduces Phosphorylated Tau Levels in Alzheimer's Disease Cell Culture Models.
Journal of Alzheimer's disease : JAD (2020-06-23)
Katelyn Cuttler, Monique J Bignoux, Tyrone C Otgaar, Stephanie Chigumba, Eloise Ferreira, Stefan F T Weiss
ABSTRACT
Alzheimer's disease (AD) is characterized by amyloid-β (Aβ) plaque and neurofibrillary tangle formation, respectively. Neurofibrillary tangles form as a result of the intracellular accumulation of hyperphosphorylated tau. Telomerase activity and levels of the human reverse transcriptase (hTERT) subunit of telomerase are significantly decreased in AD. Recently, it has been demonstrated that the 37 kDa/67 kDa laminin receptor (LRP/LR) interacts with telomerase and is implicated in Aβ pathology. Since both LRP/LR and telomerase are known to play a role in the Aβ facet of AD, we hypothesized that they might also play a role in tauopathy. This study aimed to determine if LRP/LR has a relationship with tau and whether overexpression of LRP::FLAG has an effect on tauopathy-related proteins. We employed confocal microscopy and FRET to determine whether LRP/LR and tau co-localize and interact. LRP::FLAG overexpression in HEK-293 and SH-SY5Y cells as well as analysis of tauopathy-related proteins was assessed by western blotting. We demonstrate that LRP/LR co-localizes with tau in the perinuclear cell compartment and confirmed a direct interaction between LRP/LR and tau in HEK-293 cells. Overexpression of LRP::FLAG in HEK-293 and SH-SY5Y cells decreased total and phosphorylated tau levels with a concomitant decrease in PrPc levels, a tauopathy-related protein. LRP::FLAG overexpression also resulted in increased hTERT levels. This data suggest that LRP/LR extends its role in AD through a direct interaction with tau, and recommend LRP::FLAG as a possible alternative AD therapeutic via decreasing phosphorylated tau levels.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Cy3 antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous solution (Supplied as a solution in 10 mM sodium phosphate)