MilliporeSigma
  • The Unusual Transmembrane Partition of the Hexameric Channel of the Hepatitis C Virus.
The Unusual Transmembrane Partition of the Hexameric Channel of the Hepatitis C Virus.
Structure (London, England : 1993) (2018-03-20)
Wen Chen, Jyoti Dev, Julija Mezhyrova, Liqiang Pan, Alessandro Piai, James J Chou
ABSTRACT
The p7 protein of the hepatitis C virus (HCV) can oligomerize in membrane to form cation channels. Previous studies showed that the channel assembly in detergent micelles adopts a unique flower-shaped oligomer, but the unusual architecture also presented problems for understanding how this viroporin resides in the membrane. Moreover, the oligomeric state of p7 remains controversial, as both hexamer and heptamer have been proposed. Here we address the above issues using p7 reconstituted in bicelles that mimic a lipid bilayer. We found, using a recently developed oligomer-labeling method, that p7 forms hexamers in the bicelles. Solvent paramagnetic relaxation enhancement analyses showed that the bilayer thickness around the HCV ion channel is substantially smaller than expected, and thus a significant portion of the previously assigned membrane-embedded region is solvent exposed. Our study provides an effective approach for characterizing the transmembrane partition of small ion channels in near lipid bilayer environment.
Product Description
Avanti
06:0 PC (DHPC), 1,2-dihexanoyl-sn-glycero-3-phosphocholine, chloroform